Abnova
Manufacturer Code:PAB15968
Catalog # 89-117-988
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of human SLC22A1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SLC22A1.
For Research Use Only
Protein Aliases: HOCT1; OCT1; oct1_cds; Organic cation transporter 1; solute carrier family 22 (organic cation transporter), member 1; Solute carrier family 22 member 1
Gene Aliases: HOCT1; OCT1; oct1_cds; SLC22A1
UniProt ID: (Human) O15245
Entrez Gene ID: (Human) 6580
Molecular Function: carbohydrate transporter cation transporter transfer/carrier protein transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.