Invitrogen
Manufacturer Code:PA542142
Catalog # PIPA542142
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC20A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gibbon ape leukemia virus receptor 2; GLVR-2; murine leukemia virus, amphotropic, receptor for; murine leukemia virus, amphotropic; receptor; Phosphate transporter 2; PiT-2; RAM-1; receptor for amphitropic viruses 1; Receptor for amphotropic viruses 1; Sodium-dependent phosphate transporter 2; Solute carrier family 20 (phosphate transporter) member 2 GLVR2 Glvr-2 MLVAR PIT-2 Ram1 pit2 hPit2; solute carrier family 20 (phosphate transporter), member 2; Solute carrier family 20 member 2; solute carrier family 20, member 2; Type III sodium-dependent phosphate transporter
Gene Aliases: Ab1-188; fi23g11; GLVR-2; GLVR2; IBGC1; IBGC3; MLVAR; MolPit2; PIT-2; PIT2; RAM-1; RAM1; SLC20A2; wu:fi23g11; zgc:152990
UniProt ID: (Human) Q08357, (Bovine) A1A4I1, (Mouse) Q80UP8, (Rat) Q63488
Entrez Gene ID: (Human) 6575, (Dog) 482838, (Bovine) 518905, (Rabbit) 100348436, (Horse) 100054432, (Guinea pig) 100726368, (Mouse) 20516, (Rat) 29502, (Zebrafish) 558346
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.