Invitrogen
Manufacturer Code:PA542448
Catalog # PIPA542448
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human SLC20A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gibbon ape leukemia virus receptor 1; GLVR-1; Leukemia virus receptor 1 homolog; Phosphate transporter 1; PiT-1; RPHO-1; Sodium-dependent phosphate transporter 1; Sodium-dependent phosphate transporter 1-like protein; Solute carrier family 20 (phosphate transporter) member 1 DKFZp686J2397 FLJ41426 GLVR1 Glvr-1 PIT1 PiT-1; solute carrier family 20 (phosphate transporter), member 1; Solute carrier family 20 member 1
Gene Aliases: Glvr-1; GLVR1; PiT-1; PIT1; SLC20A1
UniProt ID: (Human) Q8WUM9, (Rat) Q9JJP0
Entrez Gene ID: (Human) 6574, (Rabbit) 100358070, (Dog) 483066, (Bovine) 514255, (Pig) 100518065, (Horse) 100064887, (Rat) 81826
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.