Proteintech
Manufacturer Code:124231AP150UL
Catalog # 50-172-7972
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC20A1 Fusion Protein Ag3084 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gibbon ape leukemia virus receptor 1; Glvr 1 GLVR1 Phosphate transporter 1 PiT 1 PIT1 SLC20A1; GLVR-1; Leukemia virus receptor 1 homolog; Phosphate transporter 1; PiT-1; RPHO-1; Sodium-dependent phosphate transporter 1; solute carrier family 20 (phosphate transporter), member 1; Solute carrier family 20 member 1
Gene Aliases: AI607883; Glvr-1; GLVR1; PiT-1; PIT1; SLC20A1
UniProt ID: (Human) Q8WUM9, (Rat) Q9JJP0, (Mouse) Q61609
Entrez Gene ID: (Human) 6574, (Rat) 81826, (Mouse) 20515
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.