Novus Biologicals
Manufacturer Code:NBP297567AF647
Catalog # NB045010
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human SLC1A5 fragment. |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AAAT ASCT2M7VS1 ATB(0) Baboon M7 virus receptor M7V1ATBO neutral amino acid transporter B neutral amino acid transporter B(0) R16 RD114 virus receptor RD114/simian type D retrovirus receptor RDR RDRCFLJ31068 Sodium-dependent neutral amino acid transporter type 2 solute carrier family 1 (neutral amino acid transporter) member 5 Solute carrier family 1 member 5; ATB(0); Baboon M7 virus receptor; neutral amino acid transporter B; Neutral amino acid transporter B(0); RD114 virus receptor; RD114/simian type D retrovirus receptor; Sodium-dependent neutral amino acid transporter type 2; solute carrier family 1 (neutral amino acid transporter), member 5; Solute carrier family 1 member 5
Gene Aliases: AAAT; ASCT2; ATBO; M7V1; M7VS1; R16; RDR; RDRC; SLC1A5
UniProt ID: (Human) Q15758
Entrez Gene ID: (Human) 6510
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.