Invitrogen
Manufacturer Code:PA569260
Catalog # PIPA569260
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the C terminal region of human SLC1A4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alanine/serine/cysteine/threonine transporter 1; ASCT-1; ASCT1 SATT ASCT-1 alanine/serine/cysteine/threonine transporter 1 glutamate/neutral amino acid transporter neutral amino acid transporter A solute carrier family 1 member 4; glutamate/neutral amino acid transporter; Neutral amino acid transporter A; neutral amino acid transporter ASCT1; SATT; solute carrier family 1 (glutamate/neutral amino acid transporter), member 4; Solute carrier family 1 member 4
Gene Aliases: ASCT-1; ASCT1; AW045657; SATT; si:zc101n13.5; SLC1A4; SPATCCM; zgc:92817
UniProt ID: (Human) P43007, (Bovine) A2VDL4, (Mouse) O35874
Entrez Gene ID: (Rabbit) 100348633, (Human) 6509, (Bovine) 326577, (Horse) 100063163, (Dog) 481391, (Rat) 305540, (Guinea pig) 100724004, (Mouse) 55963, (Zebrafish) 368885
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.