Novus Biologicals
Manufacturer Code:NBP29519620UL
Catalog # NB024555
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human SLC1A4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alanine/serine/cysteine/threonine transporter 1; Alanine/serine/cysteine/threonine transporter 1 ASCT1 ASCT1 ASCT-1 neutral amino acid transporter A SATTglutamate/neutral amino acid transporter solute carrier family 1 (glutamate/neutral amino acid transporter) member 4 Solute carrier family 1 member 4; ASCT-1; glutamate/neutral amino acid transporter; Neutral amino acid transporter A; SATT; solute carrier family 1 (glutamate/neutral amino acid transporter), member 4; Solute carrier family 1 member 4
Gene Aliases: ASCT1; SATT; SLC1A4; SPATCCM
UniProt ID: (Human) P43007
Entrez Gene ID: (Human) 6509
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.