Abnova
Manufacturer Code:PAB28378
Catalog # 89-129-882
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to 17 amino acids at N-Terminus of human SLC1A4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -80°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyconal antibody raised against synthetic peptide of SLC1A4.
For Research Use Only
Protein Aliases: Alanine/serine/cysteine/threonine transporter 1; ASCT-1; ASCT1; glutamate/neutral amino acid transporter; Neutral amino acid transporter A; SATT; solute carrier family 1 (glutamate/neutral amino acid transporter), member 4; Solute carrier family 1 member 4
Gene Aliases: ASCT1; SATT; SLC1A4; SPATCCM
UniProt ID: (Human) P43007
Entrez Gene ID: (Human) 6509
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.