Invitrogen
Manufacturer Code:PA598872
Catalog # PIPA598872
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human Thiamine transporter 2 protein (191-282AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: solute carrier family 19 (thiamine transporter), member 3; Solute carrier family 19 member 3; solute carrier family 19, member 3; Thiamine transporter 2; ThTr-2
Gene Aliases: BBGD; SLC19A3; THMD2; THTR2
UniProt ID: (Human) Q9BZV2
Entrez Gene ID: (Human) 80704
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.