Invitrogen
Manufacturer Code:PA542219
Catalog # PIPA542219
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC19A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: solute carrier family 19 (sodium/hydrogen exchanger), member 3; solute carrier family 19 (thiamine transporter), member 3; Solute carrier family 19 member 3; solute carrier family 19, member 3; Thiamine transporter 2; ThTr-2
Gene Aliases: A230084E24Rik; AI788884; BBGD; SLC19A3; THMD2; THTR2
UniProt ID: (Human) Q9BZV2, (Mouse) Q99PL8
Entrez Gene ID: (Human) 80704, (Bovine) 537280, (Rabbit) 100341170, (Guinea pig) 100731585, (Rat) 316559, (Mouse) 80721
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.