Invitrogen
Manufacturer Code:PA5114216
Catalog # PIPA5114216
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Immunogen sequence: LGLEITTQFL IYASYFALIA VVFLASGAVS VMKKCRKLED PQSSSQVTTS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AV276020 AW322295 DDA1 high affinity thiamine transporter reduced folate carrier protein (RFC) like SLC19A2 solute carrier family 19 (thiamine transporter) member 2 Solute carrier family 19 member 2 TC1 Thiamine carrier 1 Thiamine transporter 1 THMD1 THT1 THTR1 ThTr-1 TRMA; high affinity thiamine transporter; reduced folate carrier protein (RFC) like; solute carrier family 19 (thiamine transporter), member 2; Solute carrier family 19 member 2; TC1; Thiamine carrier 1; Thiamine transporter 1; ThTr-1
Gene Aliases: SLC19A2; TC1; THMD1; THT1; THTR1; TRMA
UniProt ID: (Human) O60779
Entrez Gene ID: (Human) 10560
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.