Novus Biologicals
Manufacturer Code:NBP186944
Catalog # NBP186944
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:RECWFLPTALLCAYGFFASLRPSEPFLTPYLLGPDKNLTEREEPKPDRLLVLKVLWNDFLMCYSSR |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: high affinity thiamine transporter; high affinity thiamine transporter reduced folate carrier protein (RFC) like solute carrier family 19 (thiamine transporter) member 2 Solute carrier family 19 member 2 TC1 Thiamine carrier 1 thiamine transporter 1 THT1 ThTr1 ThTr-1 THTR1thTr-1 TRMA; reduced folate carrier protein (RFC) like; solute carrier family 19 (thiamine transporter), member 2; Solute carrier family 19 member 2; TC1; Thiamine carrier 1; Thiamine transporter 1; ThTr-1
Gene Aliases: SLC19A2; TC1; THMD1; THT1; THTR1; TRMA
UniProt ID: (Human) O60779
Entrez Gene ID: (Human) 10560
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.