Invitrogen
Manufacturer Code:PA566633
Catalog # PIPA566633
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human SLC18A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Chromaffin granule amine transporter; solute carrier family 18 (vesicular monoamine transporter), member 1; Solute carrier family 18 (vesicular monoamine) member 1 CGAT VAT1 VMAT1; solute carrier family 18 (vesicular monoamine), member 1; Solute carrier family 18 member 1; VAT1; Vesicular amine transporter 1
Gene Aliases: CGAT; SLC18A1; VAT1; VMAT1
UniProt ID: (Human) P54219
Entrez Gene ID: (Human) 6570
Molecular Function: amino acid transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.