Proteintech
Manufacturer Code:203401AP150UL
Catalog # 50-173-2838
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC18A1 Fusion Protein Ag14131 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CGAT SLC18A1 VAT1 Vesicular amine transporter 1 VMAT1; Chromaffin granule amine transporter; solute carrier family 18 (vesicular monoamine transporter), member 1; solute carrier family 18 (vesicular monoamine), member 1; Solute carrier family 18 member 1; solute carrier family 18, member 1; solute carrier family18 (vesicular monoamine) member 1 (chromaffin granule amine transporter); VAT1; Vesicular amine transporter 1; vesicular monoamine transporter 1; VMAT1
Gene Aliases: CGAT; SLC18A1; VAT1; VMAT-1; VMAT1
UniProt ID: (Human) P54219, (Rat) Q01818
Entrez Gene ID: (Human) 6570, (Rat) 25693
Molecular Function:
amino acid transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.