Invitrogen
Manufacturer Code:PA542456
Catalog # PIPA542456
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human SLC17A5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AST; H(+)/nitrate cotransporter; H(+)/sialic acid cotransporter; Membrane glycoprotein HP59; Membrane glycoprotein SP55; proton-coupled sialic acid transporter; sialic acid storage disease; Sialin; Sodium/sialic acid cotransporter; solute carrier family 17 (acidic sugar transporter), member 5; Solute carrier family 17 (anion/sugar transporter) member 5 AST FLJ22227 FLJ23268 ISSD NSD SD SIALIN SIASD SLD; solute carrier family 17 (anion/sugar transporter), member 5; Solute carrier family 17 member 5; Vesicular H(+)/Aspartate-glutamate cotransporter
Gene Aliases: 4631416G20Rik; 4732491M05; AST; ISSD; NSD; sb:cb809; SD; SIALIN; SIASD; SLC17A5; SLD; SP55; zgc:153077
UniProt ID: (Human) Q9NRA2, (Sheep) Q9MZD1, (Mouse) Q8BN82
Entrez Gene ID: (Dog) 474969, (Human) 26503, (Horse) 100068130, (Bovine) 530164, (Sheep) 443098, (Rabbit) 100328660, (Mouse) 235504, (Rat) 363103, (Guinea pig) 100714527, (Zebrafish) 767760
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.