Invitrogen
Manufacturer Code:PA5111910
Catalog # PIPA5111910
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide corresponding to amino acid residues 479-492 of mouse SLC17A5 (Intracellular C-terminus) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 4631416G20Rik 4732491M05 AST BOS_9478 FLJ22227 FLJ23268 H(+)/nitrate cotransporter H(+)/sialic acid cotransporter ISSD Membrane glycoprotein HP59 Membrane glycoprotein SP55 NSD sb:cb809 SD Sialin SIASD SLC17A5 SLD Sodium/sialic acid cotransporter Solute carrier family 17 member 5 SP55 Vesicular H(+)/Aspartate-glutamate cotransporter zgc:153077; AST; H(+)/nitrate cotransporter; H(+)/sialic acid cotransporter; Membrane glycoprotein HP59; proton-coupled sialic acid transporter; sialic acid storage disease; Sialin; sodium/sialic acid cotransporter; solute carrier family 17 (acidic sugar transporter), member 5; solute carrier family 17 (anion/sugar transporter), member 5; Solute carrier family 17 member 5; Vesicular H(+)/Aspartate-glutamate cotransporter
Gene Aliases: 4631416G20Rik; 4732491M05; AST; ISSD; NSD; SD; SIALIN; SIASD; SLC17A5; SLD
UniProt ID: (Human) Q9NRA2, (Mouse) Q8BN82
Entrez Gene ID: (Human) 26503, (Mouse) 235504, (Rat) 363103
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.