Novus Biologicals
Manufacturer Code:NBP159883
Catalog # NBP159883
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLC17A4(solute carrier family 17 (sodium phosphate) member 4) The peptide sequence was selected from the middle region of SLC17A4. Peptide sequence YFCEYWLFYTIMAYTPTYISSVLQANLRDSGILSALPFVVGCICIILGGL. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: KAIA2138 KIAA2138 MGC129623 Na/PO4 cotransporter putative small intestine sodium-dependent phosphate transport protein solute carrier family 17 (sodium phosphate) member 4 Solute carrier family 17 member 4; Na/PO4 cotransporter; Probable small intestine urate exporter; putative small intestine sodium-dependent phosphate transport protein; solute carrier family 17 (sodium phosphate), member 4; Solute carrier family 17 member 4
Gene Aliases: KAIA2138; SLC17A4
UniProt ID: (Human) Q9Y2C5
Entrez Gene ID: (Human) 10050
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.