Novus Biologicals
Manufacturer Code:NBP160072
Catalog # NBP160072
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLC17A2 (solute carrier family 17 (sodium phosphate) member 2) The peptide sequence was selected from the middle region of SLC17A2. Peptide sequence VIYDDPMHHPCISVREKEHILSSLAQQPSSPGRAVPIKAMVTCLPLWAIF. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MGC138238 NPT3member 2 solute carrier family 17 (sodium phosphate) member 2; Na(+)/PI cotransporter 3; sodium phosphate transporter 3; Sodium-dependent phosphate transport protein 3; Sodium/phosphate cotransporter 3; solute carrier family 17 (vesicular glutamate transporter), member 2; Solute carrier family 17 member 2
Gene Aliases: NPT3; SLC17A2
UniProt ID: (Human) O00624
Entrez Gene ID: (Human) 10246
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.