Invitrogen
Manufacturer Code:PA542458
Catalog # PIPA542458
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC16A8 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MCT 3; Monocarboxylate transporter 3; preferentially expressed in RPE cells and choroid plexus epithelium; Proton-coupled monocarboxylate transporter 3; solute carrier 16 (monocarboxylic acid transporters), member 8; solute carrier family 16 (monocarboxylate transporter), member 8; solute carrier family 16 (monocarboxylic acid transporters), member 8; Solute carrier family 16 member 8; Solute carrier family 16 member 8 (monocarboxylic acid transporter 3) MCT3 REMP; solute carrier family 16, member 8 (monocarboxylic acid transporter 3)
Gene Aliases: MCT3; REMP; SLC16A8
UniProt ID: (Human) O95907, (Mouse) O35308
Entrez Gene ID: (Horse) 100069977, (Human) 23539, (Dog) 610184, (Mouse) 57274, (Rat) 65200, (Guinea pig) 100732846, (Zebrafish) 557884
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.