Abnova
Manufacturer Code:PAB16060
Catalog # 89-118-036
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of mouse Slc16a7. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Slc16a7.
For Research Use Only
Protein Aliases: 4921534N07Rik; MCT 2; Mct2; MGC143699; MGC143700; MGC143718; Monocarboxylate transporter 2; Solute carrier family 16 member 7
Gene Aliases: 4921534N07Rik; Mct2; Slc16a7
UniProt ID: (Mouse) O70451
Entrez Gene ID: (Mouse) 20503
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.