Invitrogen
Manufacturer Code:PA576688
Catalog # PIPA576688
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit full length Human SLC16A4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MCT 4; MCT 5; Monocarboxylate transporter 4; Monocarboxylate transporter 5; Monocarboxylate transporter 5 MCT 5 Monocarboxylate transporter 4 MCT 4 Solute carrier family 16 member 4 SLC16A4 MCT4 MCT5; solute carrier family 16 (monocarboxylic acid transporters), member 4; Solute carrier family 16 member 4; solute carrier family 16, member 4 (monocarboxylic acid transporter 5)
Gene Aliases: AW146050; MCT 5; MCT4; MCT5; SLC16A4
UniProt ID: (Human) O15374, (Mouse) Q8R0M8
Entrez Gene ID: (Human) 9122, (Mouse) 229699
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.