Novus Biologicals
Manufacturer Code:NBP29489620UL
Catalog # NB024300
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 250-350 of human SLC16A4 (NP_004687.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MCT 4; MCT 5; MCT 5 MCT 6 MCT6 Monocarboxylate transporter 5 Monocarboxylic acid transporter 5 Monocarboxylic acid transporter 6 MOT5 SLC16A5 solute carrier family 16 (monocarboxylic acid transporters) member 5 Solute carrier family 16 member 5; Monocarboxylate transporter 4; Monocarboxylate transporter 5; solute carrier family 16 (monocarboxylic acid transporters), member 4; Solute carrier family 16 member 4; solute carrier family 16, member 4 (monocarboxylic acid transporter 5)
Gene Aliases: MCT4; MCT5; SLC16A4
UniProt ID: (Human) O15374
Entrez Gene ID: (Human) 9122
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.