Invitrogen
Manufacturer Code:PA582084
Catalog # PIPA582084
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SLC16A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AHDS DXS128 MCT7 MCT8 MRX22 XPCT; MCT 7; MCT 8; Monocarboxylate transporter 7; Monocarboxylate transporter 8; Monocarboxylate transporter 8 (MCT 8) (X-linked PEST-containing transporter); Solute carrier family 16 member 2; solute carrier family 16, member 2 (thyroid hormone transporter); X-linked PEST-containing transporter
Gene Aliases: AHDS; AW105741; DXS128; DXS128E; MCT 7; MCT 8; MCT7; MCT8; MRX22; SLC16A2; XPCT
UniProt ID: (Human) P36021, (Mouse) O70324
Entrez Gene ID: (Human) 6567, (Mouse) 20502
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.