Invitrogen
Manufacturer Code:PA5101676
Catalog # PIPA5101676
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human SLC16A2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AHDS AW105741 DXS128 DXS128E MCT 7 MCT 8 MCT7 Mct8 monocarboxylate transporter Monocarboxylate transporter 7 monocarboxylate transporter 8 Monocarboxylate transporter 8 (MCT 8) (X-linked PEST-containing transporter) MRX22 SLC16A2 solute carrier family 16 (monocarboxylic acid transporters) member 2 solute carrier family 16 member 2 solute carrier family 16 member 2 (monocarboxylic acid transporter 8) solute carrier family 16 member 2 (thyroid hormone transporter) X-linked PEST-containing transporter Xpct; MCT 7; MCT 8; Monocarboxylate transporter 7; Monocarboxylate transporter 8; Monocarboxylate transporter 8 (MCT 8) (X-linked PEST-containing transporter); solute carrier family 16 (monocarboxylic acid transporters), member 2; Solute carrier family 16 member 2; solute carrier family 16, member 2 (monocarboxylic acid transporter 8); solute carrier family 16, member 2 (thyroid hormone transporter); X-linked PEST-containing transporter
Gene Aliases: AHDS; AW105741; DXS128; DXS128E; MCT 7; MCT 8; MCT7; MCT8; MRX22; SLC16A2; XPCT
UniProt ID: (Human) P36021, (Rat) Q8K1P8, (Mouse) O70324
Entrez Gene ID: (Human) 6567, (Rat) 259248, (Mouse) 20502
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.