Abnova
Manufacturer Code:PAB20196
Catalog # 89-121-701
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SLC16A2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Protein Aliases: MCT 7; MCT 8; Monocarboxylate transporter 7; Monocarboxylate transporter 8; Solute carrier family 16 member 2; solute carrier family 16, member 2 (thyroid hormone transporter); X-linked PEST-containing transporter
Gene Aliases: AHDS; DXS128; DXS128E; MCT 7; MCT 8; MCT7; MCT8; MRX22; SLC16A2; XPCT
UniProt ID: (Human) P36021
Entrez Gene ID: (Human) 6567
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.