Invitrogen
Manufacturer Code:PA598535
Catalog # PIPA598535
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human Monocarboxylate transporter 12 protein (431-486AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Creatine transporter 2; CRT2; MCT 12; Monocarboxylate transporter 12; monocarboxylic acid transporter 12; Solute carrier family 16 member 12; solute carrier family 16, member 12 (monocarboxylic acid transporter 12)
Gene Aliases: CJMG; CRT2; MCT12; SLC16A12
UniProt ID: (Human) Q6ZSM3
Entrez Gene ID: (Human) 387700
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.