Novus Biologicals
Manufacturer Code:NBP159530
Catalog # NBP159530
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to SLC16A12(solute carrier family 16 member 12 (monocarboxylic acid transporter 12)) The peptide sequence was selected from the N terminal of SLC16A12. Peptide sequence WMIVAGCFLVTICTRAVTRCISIFFVEFQTYFTQDY |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CJMG DKFZp686E188 MCT 12 MCT12monocarboxylate transporter 12 solute carrier family 16 (monocarboxylic acid transporters) member 12 Solute carrier family 16 member 12 solute carrier family 16 member 12 (monocarboxylic acid transporter 12); Creatine transporter 2; CRT2; MCT 12; Monocarboxylate transporter 12; monocarboxylic acid transporter 12; Solute carrier family 16 member 12; solute carrier family 16, member 12 (monocarboxylic acid transporter 12)
Gene Aliases: CJMG; CRT2; MCT12; SLC16A12
UniProt ID: (Human) Q6ZSM3
Entrez Gene ID: (Human) 387700
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.