Abnova
Manufacturer Code:PAB17772
Catalog # 89-119-438
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to internal of human SLC16A12. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of SLC16A12.
For Research Use Only
Protein Aliases: CJMG; Creatine transporter 2; CRT2; DKFZp686E188; MCT 12; MCT12; Monocarboxylate transporter 12; monocarboxylic acid transporter 12; Solute carrier family 16 member 12; solute carrier family 16, member 12 (monocarboxylic acid transporter 12)
Gene Aliases: CJMG; CRT2; MCT12; SLC16A12
UniProt ID: (Human) Q6ZSM3
Entrez Gene ID: (Human) 387700
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.