Proteintech
Manufacturer Code:205531AP150UL
Catalog # 50-173-2958
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CJMG MCT 12 MCT12 Monocarboxylate transporter 12 SLC16A12; Creatine transporter 2; CRT2; MCT 12; Monocarboxylate transporter 12; monocarboxylic acid transporter 12; solute carrier family 16 (monocarboxylic acid transporters), member 12; Solute carrier family 16 member 12; solute carrier family 16, member 12 (monocarboxylic acid transporter 12)
Gene Aliases: 3230401A21; AW210596; CJMG; CRT2; MCT12; RGD1311468; SLC16A12
UniProt ID: (Human) Q6ZSM3, (Rat) D4A734, (Mouse) Q14CF9
Entrez Gene ID: (Human) 387700, (Rat) 309525, (Mouse) 240638
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.