Invitrogen
Manufacturer Code:PA524397
Catalog # PIPA524397
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 48-76 amino acids from the N-terminal region of human SLC16A11 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MCT 11; MCT11 MCT 11 monocarboxylate transporter 11 solute carrier family 16 (monocarboxylic acid transporters) member 11 solute carrier family 16 member 11; Monocarboxylate transporter 11; monocarboxylic acid transporter 11; novel RNA polymerase Rpb1 C-terminal repeat domain containing protein; solute carrier family 16 (monocarboxylic acid transporters), member 11; Solute carrier family 16 member 11; solute carrier family 16, member 11
Gene Aliases: AB041591; MCT11; MNCb-2717; SLC16A11
UniProt ID: (Human) Q8NCK7, (Mouse) Q5NC32
Entrez Gene ID: (Human) 162515, (Mouse) 216867
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.