Novus Biologicals
Manufacturer Code:NBP305049100UL
Catalog # NB096267
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 456-515 of human SLC16A10 (NP_061063.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Aromatic amino acid transporter 1; MCT 10; member 10 solute carrier family 16 member 10 (aromatic amino acid transporter); Monocarboxylate transporter 10; solute carrier family 16 (aromatic amino acid transporter), member 10; solute carrier family 16 (monocarboxylic acid transporters), member 10; Solute carrier family 16 member 10; solute carrier family 16, member 10 (aromatic amino acid transporter); T-type amino acid transporter 1
Gene Aliases: MCT10; PRO0813; SLC16A10; TAT1
UniProt ID: (Human) Q8TF71
Entrez Gene ID: (Human) 117247
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.