Abnova
Manufacturer Code:PAB20909
Catalog # 89-122-410
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human SLC16A10. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant SLC16A10.
For Research Use Only
Protein Aliases: Aromatic amino acid transporter 1; MCT 10; Monocarboxylate transporter 10; PRO0813; solute carrier family 16 (aromatic amino acid transporter), member 10; solute carrier family 16 (monocarboxylic acid transporters), member 10; Solute carrier family 16 member 10; solute carrier family 16, member 10 (aromatic amino acid transporter); T-type amino acid transporter 1; TAT1
Gene Aliases: MCT10; PRO0813; SLC16A10; TAT1
UniProt ID: (Human) Q8TF71
Entrez Gene ID: (Human) 117247
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.