Invitrogen
Manufacturer Code:PA553479
Catalog # PIPA553479
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human SLC15A4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hPHT1; Peptide transporter 4; peptide-histidine transporter 4; Peptide/histidine transporter 1; rPHT1; solute carrier family 15 (oligopeptide transporter), member 4; Solute carrier family 15 member 4; Solute carrier family 15 member 4 FP12591 PHT1 PTR4; solute carrier family 15, member 4
Gene Aliases: AA987064; AW742963; C130069N12Rik; FP12591; PHT1; PTR4; SLC15A4
UniProt ID: (Human) Q8N697, (Rat) O09014, (Mouse) Q91W98
Entrez Gene ID: (Human) 121260, (Rat) 246280, (Mouse) 100561
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.