Invitrogen
Manufacturer Code:PA542480
Catalog # PIPA542480
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC15A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: H+ coupled di/tri-peptide cotransporter 2; Kidney H(+)/peptide cotransporter; Oligopeptide transporter, kidney isoform; Peptide transporter 2; proton-dependent high affinity oligopeptide transporter PepT2; Slc15a2 variant A; Slc15a2 variant A2; solute carrier family 15 (H+/peptide transporter), member 2; solute carrier family 15 (oligopeptide transporter), member 2; Solute carrier family 15 member 2; solute carrier family 15, member 2
Gene Aliases: 8430408C16Rik; C78862; OPT-3; PEPT2; SLC15A2
UniProt ID: (Rabbit) P46029, (Human) Q16348
Entrez Gene ID: (Pig) 100048951, (Rabbit) 100009433, (Bovine) 506921, (Human) 6565, (Horse) 100070712, (Dog) 488006, (Mouse) 57738, (Rat) 60577, (Guinea pig) 100724012
Molecular Function:
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.