Abnova
Manufacturer Code:H00008170A01
Catalog # 89-105-814
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 40 a.a. &bsim 128 a.a) partial recombinant protein with GST tag.,SLC14A2 (NP_009094 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant SLC14A2.
For Research Use Only
Protein Aliases: FLJ16167; hUT-A6; HUT2; MGC119566; MGC119567; solute carrier family 14 (urea transporter), member 2; Solute carrier family 14 member 2; Urea transporter 2; Urea transporter, kidney; urea transporter-2; UT-A2; UT2; UTA; UTR
Gene Aliases: hUT-A6; HUT2; SLC14A2; UT-A2; UT2; UTA; UTR
UniProt ID: (Human) Q15849
Entrez Gene ID: (Human) 8170
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.