R&D Systems
Manufacturer Code:FAB8238T100UG
Catalog # FAB8238T100
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 888418 |
Immunogen | NS0 mouse myeloma cell line transfected with human SLC14A1 Accession # Q13336 |
Conjugate | Alexa Fluor 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Mouse MonoclonalAntibody
For Research Use Only
Protein Aliases: FLJ33745 HsT1341 HUT11 JK JKFLJ41687 RACH1 RACH2 solute carrier family 14 (urea transporter) member 1 (Kidd blood group) Solute carrier family 14 member 1 truncated urea transporter urea transporter 1 urea transporter JK glycoprotein Urea transporter erythrocyte urea transporter-B1 UT1 UT1UT-B1 UT-B1 UTE UTEblood group Kidd urea transporter; Kidd blood group; SLC14A1 JK; solute carrier family 14 (urea transporter), member 1 (Kidd blood group); Solute carrier family 14 member 1; Urea transporter 1; urea transporter JK glycoprotein; Urea transporter, erythrocyte; urea transporter-B1
Gene Aliases: HsT1341; HUT11; JK; RACH1; RACH2; SLC14A1; UT-B1; UT1; UTE
UniProt ID: (Human) B3KR62
Entrez Gene ID: (Human) 6563
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.