Invitrogen
Manufacturer Code:PA543133
Catalog # PIPA543133
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human SLC14A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Kidd blood group; SLC14A1 JK; solute carrier family 14 (urea transporter), member 1; solute carrier family 14 (urea transporter), member 1 (Kidd blood group); Solute carrier family 14 member 1; solute carrier family 14 member 1 (Kidd blood group); Urea transporter 1; Urea transporter B; urea transporter JK glycoprotein; urea transporter variant 2; Urea transporter, erythrocyte; urea transporter-B1; UT-1; UT-B
Gene Aliases: 2610507K20Rik; 3021401A05Rik; HsT1341; HUT11; JK; RACH1; RACH2; SLC14A1; UT-1; UT-B; UT-B1; UT1; Utb1; UTE
UniProt ID: (Bovine) Q5QF96, (Human) B3KR62, (Sheep) C5MK33, (Goat) D3INW7, (Mouse) Q8VHL0
Entrez Gene ID: (Bovine) 493988, (Human) 6563, (Pig) 100157861, (Sheep) 100302356, (Horse) 100068670, (Goat) 100860878, (Dog) 490470, (Mouse) 108052
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.