Invitrogen
Manufacturer Code:PA5110377
Catalog # PIPA5110377
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence within amino acids 100 to the C-terminus of human SLC14A1 (NP_001295208.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610507K20Rik 3021401A05Rik BOS_22241 HsT1341 HUT11 JK RACH1 RACH2 Slc14a1 Solute carrier family 14 member 1 Urea transporter 1 Urea transporter B Urea transporter erythrocyte UrT2 UT1 UT-1 UT11 UT3 UT-B UTB1 UT-B1 UTE; Kidd blood group; null; SLC14A1 JK; solute carrier family 14 (urea transporter), member 1 (Kidd blood group); Solute carrier family 14 member 1; Urea transporter 1; Urea transporter B; urea transporter JK glycoprotein; Urea transporter, erythrocyte; urea transporter-B1; UT-B
Gene Aliases: 2610507K20Rik; 3021401A05Rik; HsT1341; HUT11; JK; RACH1; RACH2; SLC14A1; UrT2; UT-B; UT-B1; UT1; UT11; UT3; UTB1; UTE
UniProt ID: (Human) B3KR62, (Mouse) Q8VHL0, (Rat) P97689
Entrez Gene ID: (Human) 6563, (Mouse) 108052, (Rat) 54301
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.