Invitrogen
Manufacturer Code:PA542526
Catalog # PIPA542526
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC13A5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na(+)/citrate cotransporter; Na+-coupled citrate transporter protein; NaCT; novel solute carrier family 13 (sodium-dependent dicarboxylate transporter) (Slc13a2 or 3) member; Sodium-coupled citrate transporter; Sodium-dependent citrate transporter; sodium-dependent dicarboxylate transporter; solute carrier family 13 (sodium-dependent citrate transporter), member 5; Solute carrier family 13 member 5
Gene Aliases: EIEE25; Indy; mINDY; NaC2/NaCT; NACT; SLC13A5
UniProt ID: (Human) Q86YT5, (Rat) Q8CJ44, (Mouse) Q67BT3
Entrez Gene ID: (Dog) 606984, (Horse) 100060798, (Rabbit) 100348089, (Bovine) 507000, (Human) 284111, (Rat) 266998, (Mouse) 237831, (Guinea pig) 100724751
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.