Invitrogen
Manufacturer Code:PA5113058
Catalog # PIPA5113058
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Solute carrier family 13 member 5 protein (156-204AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BOS_18736 EIEE25 Indy mIndy Na(+)/citrate cotransporter Na+-coupled citrate transporter protein NaC2/NaCT Nact novel solute carrier family 13 (sodium-dependent dicarboxylate transporter) (Slc13a2 or 3) member SLC13A5 Sodium-coupled citrate transporter Sodium-dependent citrate transporter sodium-dependent dicarboxylate transporter solute carrier family 13 (sodium-dependent citrate transporter) member 5 Solute carrier family 13 member 5 solute carrier family 13 member 5; Na(+)/citrate cotransporter; Na+-coupled citrate transporter protein; NaCT; novel solute carrier family 13 (sodium-dependent dicarboxylate transporter) (Slc13a2 or 3) member; Sodium-coupled citrate transporter; Sodium-dependent citrate transporter; sodium-dependent dicarboxylate transporter; solute carrier family 13 (sodium-dependent citrate transporter), member 5; Solute carrier family 13 member 5
Gene Aliases: EIEE25; Indy; mINDY; NaC2/NaCT; NACT; SLC13A5
UniProt ID: (Human) Q86YT5, (Rat) Q8CJ44, (Mouse) Q67BT3
Entrez Gene ID: (Human) 284111, (Rat) 266998, (Mouse) 237831
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.