Novus Biologicals
Manufacturer Code:NBP192394
Catalog # NBP192394
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:SQKPKFNFRSQTEEERKTPFYPPPLLDWKVTQEKVPW |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686E17257 Na(+)/citrate cotransporter NaCT NACTMGC138356 Sodium-coupled citrate transporter Sodium-dependent citrate transporter solute carrier family 13 (sodium-dependent citrate transporter) member 5 solute carrier family 13 member 5 solute carrier family 13 member 5; Na(+)/citrate cotransporter; Na+-coupled citrate transporter protein; NaCT; Sodium-coupled citrate transporter; Sodium-dependent citrate transporter; sodium-dependent dicarboxylate transporter; solute carrier family 13 (sodium-dependent citrate transporter), member 5; Solute carrier family 13 member 5
Gene Aliases: EIEE25; mIndy; NACT; SLC13A5
UniProt ID: (Human) Q86YT5
Entrez Gene ID: (Human) 284111
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.