Invitrogen
Manufacturer Code:PA5113517
Catalog # PIPA5113517
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Solute carrier family 13 member 4 protein (134-273AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9630060C05Rik BOS_4549 Na(+)/sulfate cotransporter SUT-1 NaS2 SLC13A4 sodium sulfate cotransporter-2 solute carrier family 13 (sodium/sulfate symporter) member 4 solute carrier family 13 (sodium/sulfate symporters) member 4 solute carrier family 13 (sodium/sulphate symporters) member 4 Solute carrier family 13 member 4 sulphate transporter 1 SUT1 SUT-1; Na(+)/sulfate cotransporter SUT-1; NaS2; solute carrier family 13 (sodium/sulfate symporter), member 4; solute carrier family 13 (sodium/sulfate symporters), member 4; solute carrier family 13 (sodium/sulphate symporters), member 4; Solute carrier family 13 member 4; sulphate transporter 1
Gene Aliases: NAS2; SLC13A4; SUT-1; SUT1
UniProt ID: (Human) Q9UKG4
Entrez Gene ID: (Human) 26266
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.