Invitrogen
Manufacturer Code:PA569146
Catalog # PIPA569146
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the N-terminal region of human SLC13A3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hNaDC3; mNaDC3; Na(+)/dicarboxylate cotransporter 3; NaDC-3; rNaDC3; sodium-dependent high affinity dicarboxylate transporter 3; Sodium-dependent high-affinity dicarboxylate transporter 2; sodium-dependent high-affinity dicarboxylate transporter 3; solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 3; Solute carrier family 13 member 3
Gene Aliases: NaDC-3; NADC3; SDCT2; SLC13A3
UniProt ID: (Human) Q8WWT9, (Mouse) Q91Y63
Entrez Gene ID: (Human) 64849, (Bovine) 526291, (Rabbit) 100355238, (Dog) 485910, (Rat) 64846, (Mouse) 114644, (Guinea pig) 100730446
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.