Invitrogen
Manufacturer Code:PA542433
Catalog # PIPA542433
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human SLC13A2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: intestinal sodium/dicarboxylate cotransporter; Na(+)-coupled citrate transporter; Na(+)/dicarboxylate cotransporter 1; NaDC-1; Renal sodium/dicarboxylate cotransporter; sodium-dependent dicarboxylate transporter; sodium/dicarboxylate cotransporter; solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 2; Solute carrier family 13 member 2; solute carrier family 13, member 2
Gene Aliases: INDY; mucin; NaCT; NaDC-1; NADC1; SDCT1; SLC13A2
UniProt ID: (Rabbit) Q28615, (Human) Q13183
Entrez Gene ID: (Rabbit) 100009593, (Dog) 491167, (Human) 9058, (Bovine) 505069, (Pig) 100512737, (Rat) 65202, (Guinea pig) 100726937
Molecular Function: cation transporter transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.