Novus Biologicals
Manufacturer Code:NBP182604
Catalog # NBP182604
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:ATSAMMVPIAHAVLDQLHSSQASSNVEEGSNNPTFELQEPSPQKEVTKLDNGQALPVTSASSEGRAHLSQKHLHLTQCMSLCVCYSASIGGIATLTGTAPNLVLQGQINSLFPQNGNVV |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na(+)-coupled citrate transporter; Na(+)/dicarboxylate cotransporter 1; Na(+)/dicarboxylate cotransporter 1 NADC1Na(+)-coupled citrate transporter NaDC-1NaCT Renal sodium/dicarboxylate cotransporter SDCT1 solute carrier family 13 (sodium-dependent dicarboxylate transporter) member 2 solute carrier family 13 member 2; NaDC-1; Renal sodium/dicarboxylate cotransporter; solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 2; Solute carrier family 13 member 2
Gene Aliases: NaCT; NaDC-1; NADC1; SDCT1; SLC13A2
UniProt ID: (Human) Q13183
Entrez Gene ID: (Human) 9058
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.