Proteintech
Manufacturer Code:217221AP150UL
Catalog # 50-173-3565
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | SLC13A2 Fusion Protein Ag16379 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: intestinal sodium/dicarboxylate cotransporter; Na(+)-coupled citrate transporter; Na(+)/dicarboxylate cotransporter 1; NaCT NaDC 1 NADC1 SDCT1 SLC13A2; NaDC-1; Renal sodium/dicarboxylate cotransporter; sodium-dependent dicarboxylate transporter; sodium/dicarboxylate co-transporter; solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 2; Solute carrier family 13 member 2; solute carrier family 13, member 2
Gene Aliases: mNaDC-1; mucin; NaCT; NaDC-1; NADC1; SDCT1; SLC13A2
UniProt ID: (Human) Q13183, (Mouse) Q9ES88
Entrez Gene ID: (Human) 9058, (Rat) 65202, (Mouse) 20500
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.