Novus Biologicals
Manufacturer Code:NBP2857350.1ML
Catalog # NB033664
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of Human SLC13A1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na(+)/sulfate cotransporter; Renal sodium/sulfate cotransporter; solute carrier family 13 (sodium/sulfate symporters), member 1; solute carrier family 13 (sodium/sulphate symporters), member 1; Solute carrier family 13 member 1; solute carrier family 13 member 1 NAS1 NaSi-1 solute carrier family 13 (sodium/sulfate symporters) member 1
Gene Aliases: NAS1; NaSi-1; NASI1; SLC13A1
UniProt ID: (Human) Q9BZW2
Entrez Gene ID: (Human) 6561
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.