Invitrogen
Manufacturer Code:PA580005
Catalog # PIPA580005
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human SLC12A6. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Electroneutral potassium-chloride cotransporter 3; furosemide-sensitive KCl cotransporter 3; K-Cl cotransporter 3; potassium chloride cotransporter 3; potassium chloride cotransporter KCC3a-S3; potassium-chloride transporter-3a; potassium-chloride transporter-3b; solute carrier family 12 (potassium/chloride transporter), member 6; solute carrier family 12 (potassium/chloride transporters), member 6; Solute carrier family 12 member 6; Solute carrier family 12 member 6 Electroneutral potassium-chloride cotransporter 3 K-Cl cotransporter 3 SLC12A6 KCC3
Gene Aliases: 9530023I19Rik; ACCPN; gaxp; KCC3; KCC3A; KCC3B; SLC12A6
UniProt ID: (Human) Q9UHW9, (Mouse) Q924N4
Entrez Gene ID: (Human) 9990, (Mouse) 107723, (Rat) 691209
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.