Invitrogen
Manufacturer Code:PA580004
Catalog # PIPA580004
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived human SLC12A3 recombit protein (Position: H682-D936). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na-Cl cotransporter; Na-Cl symporter; NaCl electroneutral thiazide-sensitive cotransporter; NCC; solute carrier family 12 (sodium/chloride transporter), member 3; solute carrier family 12 (sodium/chloride transporters), member 3; Solute carrier family 12 member 3; Solute carrier family 12 member 3 Na-Cl cotransporter NCC Na-Cl symporter Thiazide-sensitive sodium-chloride cotransporter SLC12A3 NCC TSC; solute carrier family 12, member 3; thiazide-sensitive Na-Cl cotransporter; Thiazide-sensitive sodium-chloride cotransporter
Gene Aliases: AI035291; NCC; NCCT; SLC12A3; TSC
UniProt ID: (Human) P55017, (Rat) P55018, (Mouse) P59158
Entrez Gene ID: (Human) 6559, (Rat) 54300, (Mouse) 20497
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.