Invitrogen
Manufacturer Code:PA577816
Catalog # PIPA577816
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunomicroscopy (IM) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced against a synthetic peptide mapping to a segment of rat NCC (amino acids 74-95) N-terminal |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Na-Cl cotransporter; Na-Cl symporter; NaCl electroneutral thiazide-sensitive cotransporter; NCC; SLC12A3 SCYL1CKb10 MCP-4 MGC17134 NCC-1 NCC1 SCYA13 CK-beta-10 monocyte chemoattractant protein 4 monocyte chemotactic protein 4 new CC chemokine 1 small inducible cytokine A13 small inducible cytokine subfamily A (Cys-Cys) member 13 chemokine (C-C); solute carrier family 12 (sodium/chloride transporter), member 3; solute carrier family 12 (sodium/chloride transporters), member 3; Solute carrier family 12 member 3; solute carrier family 12, member 3; thiazide-sensitive Na-Cl cotransporter; Thiazide-sensitive sodium-chloride cotransporter
Gene Aliases: AI035291; NCC; NCCT; SLC12A3; TSC
UniProt ID: (Human) P55017, (Rat) P55018, (Mouse) P59158
Entrez Gene ID: (Human) 6559, (Dog) 478118, (Rat) 54300, (Mouse) 20497
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.